Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IL1RAPL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109853
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109853 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109853 Supplier Invitrogen™ Supplier No. PA5109853
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RQHFGADETN DDNKEYDAYL SYTKVDQDTL DCDNPEEEQF ALEVLPDVLE KHYGYKLFIP ERDLIPSGTY MEDLTRYVEQ SRRLIIVLTP DYILRRGWSI FELESRLHNM LVSGEIKVIL IECTELKGKV NCQEVESLKR SIKLLSLIKW KGSKSSKLNS KFWKHLVYEM PIKKKEMLPR CHVLDSAEQG LFGELQPIPS IAMTSTSATL VSSQADLPEF HPSDSMQIRH CCRGYKHEIP ATTLPVPSLG NHHTYCNLPL TLLNGQLPLN NTLKDTQEFH RNSSLLPLSS KELSFTSDIW.

The protein encoded by this gene is a member of the interleukin 1 receptor family. This protein is similar to the interleukin 1 accessory proteins, and is most closely related to interleukin 1 receptor accessory protein-like 1 (IL1RAPL1). This gene and IL1RAPL1 are located at a region on chromosome X that is associated with X-linked non-syndromic mental retardation. [provided by RefSeq, Jul 2008].
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL1RAPL2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.01 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene IL1RAPL2
Gene Accession No. Q9NP60
Gene Alias IL-1 receptor accessory protein-like 2; IL1R9; IL-1R9; IL-1R-9; IL1RAPL2; IL-1-RAPL-2; IL-1RAPL-2; IL1RAPL-2; IL1RAPL-2-related protein; interleukin 1 receptor 9; interleukin 1 receptor accessory protein like 2; interleukin 1 receptor accessory protein-like 2; Interleukin-1 receptor 9; RGD1561761; three immunoglobulin domain-containing IL-1 receptor-related 1; TIGIRR-1; X-linked interleukin-1 receptor accessory protein-like 2
Gene Symbols IL1RAPL2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 387-686 of human IL1RAPL2 (NP_059112.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26280
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.