Learn More
Invitrogen™ IL1RL2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA587629
Description
Immunogen sequence: MWSLLLCGLS IALPLSVTAD GCKDIFMKNE ILSASQPFAF NCTFPPITSG EVSVTWYKNS SKIPVSKIIQ SRIHQDETWI LFLPMEWGDS GVYQCVIKGR DSCHRIHVNL TVFEKH; Positive Samples: MCF7, SW620, K-562, Mouse lung, Mouse heart; Cellular Location: Membrane, Single-pass type I membrane protein.
The protein encoded by this gene is a member of the interleukin 1 receptor family. An experiment with transient gene expression demonstrated that this receptor was incapable of binding to interleukin 1 alpha and interleukin 1 beta with high affinity. This gene and four other interleukin 1 receptor family genes, including interleukin 1 receptor, type I (IL1R1), interleukin 1 receptor, type II (IL1R2), interleukin 1 receptor-like 1 (IL1RL1), and interleukin 18 receptor 1 (IL18R1), form a cytokine receptor gene cluster in a region mapped to chromosome 2q12.
Specifications
IL1RL2 | |
Polyclonal | |
Unconjugated | |
IL1RL2 | |
AI481289; AW551444; IL-1 receptor-related protein 2; IL1RL2; IL1RRP2; IL-1Rrp2; IL1R-rp2; IL-36 receptor; IL-36R; interleukin 1 receptor like 2; interleukin 1 receptor-like 2; interleukin-1 receptor related protein 2; interleukin-1 receptor-like 2; Interleukin-1 receptor-related protein 2 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
107527, 8808 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot, Immunocytochemistry | |
1.28 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9ERS7, Q9HB29 | |
IL1RL2 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-116 of human IL36R (NP_0038452). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.