Learn More
IL2 Mouse anti-Human, Clone: OTI2C9, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF810724

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Interleukin 2 (IL-2) is an immuno-modulatory cytokine that is important for the proliferation of activated T cells, differentiation of B cells, natural killer cells, monocytes and macrophages. IL-2 signals through the IL-2 receptor (IL-2R), a heterotrimeric protein complex whose gamma chain is also shared by interleukin 4 (IL-4) and interleukin 7 (IL-7). The expression of the IL-2 gene in mature thymocytes is monoallelic, which represents an unusual regulatory mode for controlling the precise expression of a single gene. The targeted disruption of a gene similar to IL-2 in mice leads to an ulcerative colitis-like disease that suggests an essential role of this gene in the immune response to antigenic stimuli.Specifications
IL2 | |
Monoclonal | |
Unconjugated | |
IL2 | |
IL-2, TCGF, lymphokine | |
Mouse | |
Affinity Chromatography | |
RUO | |
3558 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI2C9 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P60568 | |
IL2 | |
Human recombit protein fragment corresponding to amino acids 21-153 of human IL2 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.