Learn More
IL20RA Mouse anti-Human, Clone: OTI6F8, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF811647

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The protein encoded by this gene is a receptor for interleukin 20 (IL20), a cytokine that may be involved in epidermal function. The receptor of IL20 is a heterodimeric receptor complex consisting of this protein and interleukin 20 receptor beta (IL20B). This gene and IL20B are highly expressed in skin. The expression of both genes is found to be upregulated in Psoriasis.Specifications
IL20RA | |
Monoclonal | |
Unconjugated | |
IL20RA | |
CRF2-8, IL-20R-alpha, IL-20R1, IL-20RA | |
Mouse | |
Affinity Chromatography | |
RUO | |
53832 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI6F8 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9UHF4 | |
IL20RA | |
Human recombit protein fragment corresponding to amino acids 272-553 of human IL20RA produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.