Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ IL20RA Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA559935
Description
Immunogen sequence: DYEHQYYAKV KAIWGTKCSK WAESGRFYPF LETQIGPPEV ALTTDEKSIS VVLTAPEKWK RNPEDLPVSM QQIYSNLKYN VSVLNTKSNR TWSQCVTNHT LVLTWLEPNT LYCVHVESFV PGPPRRAQPS EKQCARTLK Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 76%.
The protein encoded by this gene is a receptor for interleukin 20 (IL20), a cytokine that may be involved in epidermal function. The receptor of IL20 is a heterodimeric receptor complex consisting of this protein and interleukin 20 receptor beta (IL20B). This gene and IL20B are highly expressed in skin. The expression of both genes is found to be upregulated in Psoriasis.
Specifications
IL20RA | |
Polyclonal | |
Unconjugated | |
Il20ra | |
class II cytokine receptor ZCYTOR7; CRF2-8; Cytokine receptor class-II member 8; Cytokine receptor family 2 member 8; E230031K19; IL-20 receptor subunit alpha; IL-20R1; IL20RA; IL-20RA; IL-20R-alpha; interleukin 20 receptor alpha subunit; interleukin 20 receptor subunit alpha; interleukin 20 receptor, alpha; interleukin-20 receptor I; interleukin-20 receptor subunit alpha; PRO1315; UNQ681; UNQ681/PRO1315; ZcytoR7 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
53832 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9UHF4 | |
Il20ra | |
Recombinant protein corresponding to Human IL20RA. Recombinant protein control fragment (Product #RP-89983). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction