Learn More
Invitrogen™ IL20RA Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA587796
Description
Positive Samples: MCF-7, U-87MG, A-375, SW620, Mouse liver; Cellular Location: Membrane, Single-pass type I membrane protein Immunogen sequence: VPCVSGGLPK PANITFLSIN MKNVLQWTPP EGLQGVKVTY TVQYFIYGQK KWLNKSECRN INRTYCDLSA ETSDYEHQYY AKVKAIWGTK CSKWAESGRF YPFLETQIGP PEVALTTDEK SISVVLTAPE KWKRNPEDLP VSMQQIYSNL KYNVSVLNTK SNRTWSQCVT NHTLVLTWLE PNTLYCVHVE SFVPGPPRRA QPSEKQCART LKDQSSEFKA K.
The protein encoded by this gene is a receptor for interleukin 20 (IL20), a cytokine that may be involved in epidermal function. The receptor of IL20 is a heterodimeric receptor complex consisting of this protein and interleukin 20 receptor beta (IL20B). This gene and IL20B are highly expressed in skin. The expression of both genes is found to be upregulated in Psoriasis.
Specifications
IL20RA | |
Polyclonal | |
Unconjugated | |
Il20ra | |
class II cytokine receptor ZCYTOR7; CRF2-8; Cytokine receptor class-II member 8; Cytokine receptor family 2 member 8; E230031K19; IL-20 receptor subunit alpha; IL-20R1; IL20RA; IL-20RA; IL-20R-alpha; interleukin 20 receptor alpha subunit; interleukin 20 receptor subunit alpha; interleukin 20 receptor, alpha; interleukin-20 receptor I; interleukin-20 receptor subunit alpha; PRO1315; UNQ681; UNQ681/PRO1315; ZcytoR7 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
237313, 53832 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.58 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q6PHB0, Q9UHF4 | |
Il20ra | |
Recombinant fusion protein containing a sequence corresponding to amino acids 30-250 of human IL20RA (Q9UHF4). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.