Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IL20RA Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5119916
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5119916 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5119916 Supplier Invitrogen™ Supplier No. PA5119916
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive Control: MCF-7 cell lysate, SH-SY5Y cell lysate, SH-SY5Y, human kidney tissue. Subcellular Location: Membrane.

The protein encoded by this gene is a receptor for interleukin 20 (IL20), a cytokine that may be involved in epidermal function. The receptor of IL20 is a heterodimeric receptor complex consisting of this protein and interleukin 20 receptor beta (IL20B). This gene and IL20B are highly expressed in skin. The expression of both genes is found to be upregulated in Psoriasis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL20RA
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol, 0.05% BSA and 0.05% sodium azide; pH 7.4
Gene Il20ra
Gene Accession No. Q9UHF4
Gene Alias class II cytokine receptor ZCYTOR7; CRF2-8; Cytokine receptor class-II member 8; Cytokine receptor family 2 member 8; E230031K19; IL-20 receptor subunit alpha; IL-20R1; IL20RA; IL-20RA; IL-20R-alpha; interleukin 20 receptor alpha subunit; interleukin 20 receptor subunit alpha; interleukin 20 receptor, alpha; interleukin-20 receptor I; interleukin-20 receptor subunit alpha; PRO1315; UNQ681; UNQ681/PRO1315; ZcytoR7
Gene Symbols Il20ra
Host Species Rabbit
Immunogen Synthetic peptide within human IL-20R1 aa 471-520.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 53832
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.