Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL20RA Recombinant Rabbit Monoclonal Antibody (038)
IL20RA Recombinant Rabbit Monoclonal Antibody (38)
Supplier: Thermo Scientific MA529356
Description
IL20RA Recombinant Monoclonal Antibody for IHC (P), ELISA

Specifications
IL20RA | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
Q9UHF4 | |
Il20ra | |
Recombinant Human IL20RA/IL-20RA protein (Met1-Lys250). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
ELISA, Immunohistochemistry (Paraffin) | |
038 | |
Unconjugated | |
Il20ra | |
class II cytokine receptor ZCYTOR7; CRF2-8; Cytokine receptor class-II member 8; Cytokine receptor family 2 member 8; E230031K19; IL-20 receptor subunit alpha; IL-20R1; IL20RA; IL-20RA; IL-20R-alpha; interleukin 20 receptor alpha subunit; interleukin 20 receptor subunit alpha; interleukin 20 receptor, alpha; interleukin-20 receptor I; interleukin-20 receptor subunit alpha; PRO1315; UNQ681; UNQ681/PRO1315; ZcytoR7 | |
Rabbit | |
Protein A | |
RUO | |
53832 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction