Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IL20RB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570502
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570502 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570502 Supplier Invitrogen™ Supplier No. PA570502
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 85%; Dog: 86%; Horse: 86%; Human: 100%; Mouse: 92%; Pig: 93%; Rabbit: 86%; Rat: 77%.

IL20RB and IL20RA (MIM 605620) form a heterodimeric receptor for interleukin-20 (IL20; MIM 605619) (Blumberg et al., 2001 [PubMed 11163236]).[supplied by OMIM, Feb 2009].
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL20RB
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Il20rb
Gene Accession No. 0
Gene Alias AV228068; DIRS1; Fibronectin type III domain containing 6; FNDC6; Gm186; IL-20 receptor subunit beta; IL20R2; IL-20R2; IL20RB; IL-20RB; IL-20R-beta; interleukin 20 receptor beta; interleukin 20 receptor beta subunit; interleukin 20 receptor subunit beta; interleukin-20 receptor II; interleukin-20 receptor subunit beta; RGD1566151; UNQ557/PRO1114
Gene Symbols Il20rb
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of mouse IL20RB.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 213208
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.