Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IL21R Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596690
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596690 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596690 Supplier Invitrogen™ Supplier No. PA596690
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GVSMPRGWAA PLLLLLLQGG WGCPDLVCYT DYLQTVICIL EMWNLHPSTL TLTWQDQYEE LKDEATSCSL HRSAHNATHA TYTCHMDVFH FMADDIFSVN ITDQSGNYSQ ECGSFLLAES IKPAPPFNVT VTFSGQYNIS WRSDYEDPAF YMLKGKLQYE LQYRNRGDPW AVSPRRKLIS VDSRSVSLLP LEFRKDSSYE LQVRAGPMPG SSYQGTWSEW S; Positive Samples: U-87MG, Mouse spleen, Mouse thymus; Cellular Location: Membrane, Single-pass type I membrane protein.

The protein encoded by this gene is a cytokine receptor for interleukin 21 (IL21). It belongs to the type I cytokine receptors, and has been shown to form a heterodimeric receptor complex with the common gamma-chain, a receptor subunit also shared by the receptors for interleukin 2 (IL2) and interleukin 5 (IL5). This receptor transduces the growth promoting signal of IL21, and is important for the proliferation and differentiation of T cells, B cells, and natural killer (NK) cells. The ligand binding of this receptor leads to the activation of multiple downstream signaling molecules, including JAK1, JAK3, STAT1, and STAT3. Knockout studies of a similar gene in mouse suggest a role for this gene in regulating immunoglobulin production. Three alternatively spliced transcript variants encoding the same protein have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL21R
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.21 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Il21r
Gene Accession No. Q9HBE5, Q9JHX3
Gene Alias CD360; IL-21 receptor; Il21r; IL-21R; interleukin 21 receptor; interleukin-21 receptor; LR-beta; lymphocyte receptor beta; MGC10967; NILR; novel cytokine receptor 8; Novel interleukin receptor; NR8; UNQ3121/PRO10273
Gene Symbols Il21r
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 20-240 of human IL21R (NP_8515654).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 50615, 60504
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.