Learn More
Invitrogen™ IL21R Polyclonal Antibody

Description
Positive Control: Rat spleen tissue lysate, rat liver tissue lysate, rat thymus tissue lysate, mouse spleen tissue lysate, mouse thymus tissue lysate, Daudi cell lysate, human spleen tissue. Subcellular Location: Membrane.
Specifications
Specifications
| Antigen | IL21R |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | Il21r |
| Gene Accession No. | Q9HBE5, Q9JHX3 |
| Gene Alias | CD360; IL-21 receptor; Il21r; IL-21R; interleukin 21 receptor; interleukin-21 receptor; LR-beta; lymphocyte receptor beta; MGC10967; NILR; novel cytokine receptor 8; Novel interleukin receptor; NR8; UNQ3121/PRO10273 |
| Gene Symbols | Il21r |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.