Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ IL22RA2 Monoclonal Antibody (9A11E4F7)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA530574
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human IL22RA2/IL22BP.
IL22, a homolog of IL10, is secreted by T cells and binds to and signals through the class II cytokine receptor family (CRF) heterodimer IL22RA /IL10RB. IL22 induces the production of acute-phase reactants.
Specifications
IL22RA2 | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
Q969J5 | |
IL22RA2 | |
Recombinant Human IL22BP protein (Met1-Pro231). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA | |
9A11E4F7 | |
Unconjugated | |
IL22RA2 | |
CRF2-10; CRF2-S1; CRF2X; Cytokine receptor class-II member 10; Cytokine receptor family 2 member 10; cytokine receptor family II soluble 1; cytokine receptor family type 2, soluble 1; IL-22 receptor subunit alpha-2; IL22BP; IL-22BP; Il22ra2; IL-22RA2; IL-22R-alpha-2; interleukin 22 receptor subunit alpha 2; interleukin 22 receptor, alpha 2; interleukin 22-binding protein; interleukin-22 receptor subunit alpha-2; Interleukin-22-binding protein; UNQ5793/PRO19598/PRO19822; ZcytoR16 | |
Mouse | |
Protein A | |
RUO | |
116379 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction