Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL22RA2 Mouse anti-Human, Clone: 5G11A9, Proteintech
SDP

Catalog No. 501736624 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-6624 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-6624 Supplier Proteintech Group Inc Supplier No. 661901IG150UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IL22, a homolog of IL10, is secreted by T cells and binds to and signals through the class II cytokine receptor family (CRF) heterodimer IL22RA /IL10RB. IL22 induces the production of acute-phase reactants.

Specifications

Antigen IL22RA2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 5G11A9
Concentration 1.35 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene IL22RA2
Gene Accession No. Q969J5
Gene Alias CRF2 10, CRF2 S1, CRF2X, IL 22 receptor subunit alpha 2, IL 22BP, IL 22R alpha 2, IL 22RA2, IL22BP, IL22RA2, Interleukin 22 binding protein, ZcytoR16
Gene Symbols IL22RA2
Host Species Mouse
Immunogen IL22RA2 Fusion Protein Ag18682
Purification Method Protein A
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116379
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.