Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IL22RA2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA521490
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA521490 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA521490 Supplier Invitrogen™ Supplier No. PA521490
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: HeLaS3. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

IL22, a homolog of IL10, is secreted by T cells and binds to and signals through the class II cytokine receptor family (CRF) heterodimer IL22RA /IL10RB. IL22 induces the production of acute-phase reactants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL22RA2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.66 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene IL22RA2
Gene Accession No. Q969J5
Gene Alias CRF2-10; CRF2-S1; CRF2X; Cytokine receptor class-II member 10; Cytokine receptor family 2 member 10; cytokine receptor family II soluble 1; cytokine receptor family type 2, soluble 1; IL-22 receptor subunit alpha-2; IL22BP; IL-22BP; Il22ra2; IL-22RA2; IL-22R-alpha-2; interleukin 22 receptor subunit alpha 2; interleukin 22 receptor, alpha 2; interleukin 22-binding protein; interleukin-22 receptor subunit alpha-2; Interleukin-22-binding protein; UNQ5793/PRO19598/PRO19822; ZcytoR16
Gene Symbols IL22RA2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human IL22 Receptor alpha 2. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116379
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.