Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IL23R Monoclonal Antibody (258010)
GREENER_CHOICE

Catalog No. PIMA523906
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA523906 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA523906 Supplier Invitrogen™ Supplier No. MA523906
Add to Cart
Add to Cart

Rat Monoclonal Antibody

In Western blots, this antibody does not cross-react with recombinant human IL-23 R. Reconstitute at 0.5 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.

IL-23R is a 629 aa, novel subunit of the receptor for cytokine IL23A/IL23 which pairs with the receptor molecule IL12RB1/IL12R beta 1and thus take part in IL23A signaling. Signal transduction analysis confirms that this protein associates constitutively with Janus kinase 2 (JAK2), and also binds to transcription activator STAT3 in a ligand-dependent manner. Alternative splicing results in six spliced isoforms of IL-23R. Reports suggest that IL-23R plays an important role in psoriasis pathogenesis and is also a potential therapeutic target in inflammatory bowel disease.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL23R
Applications Neutralization, Western Blot
Classification Monoclonal
Clone 258010
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene Il23r
Gene Accession No. Q8K4B4
Gene Alias IBD17; IL 23R; IL-23 receptor; IL23R; IL-23R; interleukin 23 receptor; interleukin 23 receptor-like; interleukin-23 receptor
Gene Symbols Il23r
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse IL-23 R Gly24-Asp372.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 209590
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.