Learn More
Invitrogen™ IL24 Monoclonal Antibody (303308)

Description
No cross-reactivity with recombinant human IL-24 is observed. Reconstitute at 0.5 mg/mL in sterile PBS.
Specifications
Specifications
| Antigen | IL24 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 303308 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 5% trehalose and No Preservative; pH 7.4 |
| Gene | IL24 |
| Gene Accession No. | Q925S4 |
| Gene Alias | C49A; Cytokine-like protein Mob-5; FISP; IL 24; IL10B; Il24; IL-24; IL-4-induced secreted protein; Interleukin; interleukin 24; interleukin-24; Mda7; MDA-7; melanocyte-associated Mda-7; melanoma differentiation-associated gene 7 protein; Mob5; ST16; suppression of tumorigenicity 16 (melanoma differentiation); Suppression of tumorigenicity 16 protein; Th2-specific cytokine FISP |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.