Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL28A Mouse anti-Human, Clone:6H9E6, Abnova™
Mouse monoclonal antibody raised against recombinant human IL28A.
Supplier: Abnova Corporation MAB17176
Description
This gene encodes a cytokine distantly related to type I interferons and the IL-10 family. This gene, interleukin 28B (IL28B), and interleukin 29 (IL29) are three closely related cytokine genes that form a cytokine gene cluster on a chromosomal region mapped to 19q13. Expression of the cytokines encoded by the three genes can be induced by viral infection. All three cytokines have been shown to interact with a heterodimeric class II cytokine receptor that consists of interleukin 10 receptor, beta (IL10RB) and interleukin 28 receptor, alpha (IL28RA). [provided by RefSeq]Specifications
IL28A | |
Monoclonal | |
Unconjugated | |
ELISA (1:10000) Western Blot (1:500-1:2000) Immunohistochemistry Immunocytochemistry (1:200-1:1000) Flow Cytometry The optimal working dilution should be determined by the end user. |
|
IL28A | |
Mouse | |
100 μg | |
Human | |
Antibody | |
IgG1 |
ELISA, Immunocytochemistry, Western Blot | |
6H9E6 | |
interleukin 28A (interferon, lambda 2) | |
In PBS (0.05% sodium azide) | |
IFNL2, IL-28A | |
Recombinant protein corresponding to amino acid 1-200 of human IL28A from E. coli. | |
282616 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction