Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL2RA Mouse anti-Human, Unconjugated, Clone: IL2RA/2395, Abnova™

Catalog No. 89337746 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-337-746 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-337-746 Supplier Abnova Corporation Supplier No. MAB21255
Add to Cart
Add to Cart

Mouse Monoclonal Antibody.

IL2RA Monoclonal antibody specifically detects IL2RA in Human samples. It is validated for IHC-P.

Specifications

Antigen IL2RA
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone IL2RA/2395
Conjugate Unconjugated
Description interleukin 2 receptor, alpha
Gene IL2RA
Gene Alias CD25, IDDM10, IL2R, TCGFR
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 42-183 of human IL2RA
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 3559
Target Species Human
Content And Storage Store at 4°C
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.