Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL2RA Mouse anti-Human, Unconjugated, Clone: IL2RA/423, Abnova™

Catalog No. 89337588 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-337-588 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-337-588 Supplier Abnova Corporation Supplier No. MAB21095
Add to Cart
Add to Cart

Mouse Monoclonal Antibody.

IL2RA Monoclonal antibody specifically detects IL2RA in Human samples. It is validated for IF, Flow Cyt.

Specifications

Antigen IL2RA
Applications Immunofluorescence, Flow Cytometry
Classification Monoclonal
Clone IL2RA/423
Conjugate Unconjugated
Description interleukin 2 receptor, alpha
Gene IL2RA
Gene Alias CD25, IDDM10, IL2R, TCGFR
Host Species Mouse
Immunogen Recombinant protein corresponding to human IL2RA
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 3559
Target Species Human
Content And Storage Store at 2 to 8°C
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.