Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL2RB 293T Cell Overexpression Lysate (Denatured), Abnova

Catalog No. 89009152 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-009-152 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-009-152 Supplier Abnova Corporation Supplier No. H00003560T01
Add to Cart
Add to Cart

Western Blotting

Specifications

For Use With (Application) Western Blot
Cell Line 293T
Description IL2RB 293T Cell Transient Overexpression Lysate(Denatured)
Gene Accession No. NM_000878.2
Molecular Weight (g/mol) 61.1
pH Range 6.8
Plasmid pCMV-IL2RB full-length
Quality Control Testing Transient overexpression cell Lysate was tested with Anti-IL2RB antibody (H00003560-B01) by western blotting
Quantity 100 μL
Storage Buffer 1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Host Species Human
Content And Storage Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.