Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IL2RG Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597868
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597868 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597868 Supplier Invitrogen™ Supplier No. PA597868
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

IL2RG Polyclonal Antibody for Western Blot, IHC (P), ELISA

IL2RG is an important signaling component of many interleukin receptors, including those of interleukin -2, -4, -7 and -21, and is thus referred to as the common gamma chain. Mutations in this gene cause X-linked severe combined immunodeficiency (XSCID), as well as X-linked combined immunodeficiency (XCID), a less severe immunodeficiency disorder.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL2RG
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene IL2RG
Gene Accession No. P31785
Gene Alias [g]c; Ab2-183; CD132; CD132 antigen; CIDX; common cytokine receptor gamma chain; cytokine receptor common subunit gamma; gamma(c); gammaC; gc; IL-2 receptor subunit gamma; IL2R gamma; IL-2R subunit gamma; IL2RG; IL-2RG; IMD4; interleukin 2 receptor subunit gamma; interleukin 2 receptor, gamma; interleukin 2 receptor, gamma (severe combined immunodeficiency); interleukin 2 receptor, gamma chain; Interleukin-2 receptor subunit gamma; p64; SCIDX; SCIDX1
Gene Symbols IL2RG
Host Species Rabbit
Immunogen Recombinant Human Cytokine receptor common subunit gamma protein (75-230AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3561
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.