Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ IL2RG Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5115413
Description
Antibody detects endogenous levels of total IL2RG.
IL2RG is an important signaling component of many interleukin receptors, including those of interleukin -2, -4, -7 and -21, and is thus referred to as the common gamma chain. Mutations in this gene cause X-linked severe combined immunodeficiency (XSCID), as well as X-linked combined immunodeficiency (XCID), a less severe immunodeficiency disorder.
Specifications
| IL2RG | |
| Polyclonal | |
| Unconjugated | |
| IL2RG | |
| [g]c; Ab2-183; CD132; CD132 antigen; CIDX; common cytokine receptor gamma chain; cytokine receptor common subunit gamma; gamma(c); gammaC; gc; IL-2 receptor subunit gamma; IL2R gamma; IL-2R subunit gamma; IL2RG; IL-2RG; IMD4; interleukin 2 receptor subunit gamma; interleukin 2 receptor, gamma; interleukin 2 receptor, gamma (severe combined immunodeficiency); interleukin 2 receptor, gamma chain; Interleukin-2 receptor subunit gamma; p64; SCIDX; SCIDX1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 140924, 16186, 3561 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (PFA fixed), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| P31785, P34902 | |
| IL2RG | |
| A synthesized peptide derived from human IL2RG(Accession P31785), corresponding to amino acid residues S53-S103. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction