Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IL2RG Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5119907
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5119907 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5119907 Supplier Invitrogen™ Supplier No. PA5119907
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive Control: Mouse spleen tissue lysates, SH-SY5Y, SiHa, human spleen tissue, human tonsil tissue. Subcellular Location: Cell membrane, Cell surface.

IL2RG is an important signaling component of many interleukin receptors, including those of interleukin -2, -4, -7 and -21, and is thus referred to as the common gamma chain. Mutations in this gene cause X-linked severe combined immunodeficiency (XSCID), as well as X-linked combined immunodeficiency (XCID), a less severe immunodeficiency disorder.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL2RG
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene IL2RG
Gene Accession No. P31785, P34902
Gene Alias [g]c; Ab2-183; CD132; CD132 antigen; CIDX; common cytokine receptor gamma chain; cytokine receptor common subunit gamma; gamma(c); gammaC; gc; IL-2 receptor subunit gamma; IL2R gamma; IL-2R subunit gamma; IL2RG; IL-2RG; IMD4; interleukin 2 receptor subunit gamma; interleukin 2 receptor, gamma; interleukin 2 receptor, gamma (severe combined immunodeficiency); interleukin 2 receptor, gamma chain; Interleukin-2 receptor subunit gamma; p64; SCIDX; SCIDX1
Gene Symbols IL2RG
Host Species Rabbit
Immunogen Synthetic peptide within human IL-2RG aa 301-350.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16186, 3561
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.