Learn More
Invitrogen™ IL2RG Polyclonal Antibody
Goat Polyclonal Antibody
Supplier: Invitrogen™ PA547409
Description
In direct ELISAs, approximately 50% cross-reactivity with recombinant mouse IL-2 R gamma is observed, less than 10% cross-reactivity with recombinant human (rh) IL-2 R beta is observed, and less than 2% cross-reactivity with rhIL-2 R alpha is observed. Reconstitute at 0.2 mg/mL in sterile PBS. Endoxin level is <1.0 EU per 1 μg of the antibody by the LAL method.
IL2RG is an important signaling component of many interleukin receptors, including those of interleukin -2, -4, -7 and -21, and is thus referred to as the common gamma chain. Mutations in this gene cause X-linked severe combined immunodeficiency (XSCID), as well as X-linked combined immunodeficiency (XCID), a less severe immunodeficiency disorder.
Specifications
IL2RG | |
Polyclonal | |
Unconjugated | |
IL2RG | |
[g]c; Ab2-183; CD132; CD132 antigen; CIDX; common cytokine receptor gamma chain; cytokine receptor common subunit gamma; gamma(c); gammaC; gc; IL-2 receptor subunit gamma; IL2R gamma; IL-2R subunit gamma; IL2RG; IL-2RG; IMD4; interleukin 2 receptor subunit gamma; interleukin 2 receptor, gamma; interleukin 2 receptor, gamma (severe combined immunodeficiency); interleukin 2 receptor, gamma chain; Interleukin-2 receptor subunit gamma; p64; SCIDX; SCIDX1 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
3561 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Neutralization, Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
P31785 | |
IL2RG | |
Sf 21-derived recombinant human Common gamma Chain/IL-2R gamma Leu23-Asn254. | |
250 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.