Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IL36B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547066
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547066 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547066 Supplier Invitrogen™ Supplier No. PA547066
Add to Cart
Add to Cart

Goat Polyclonal Antibody

In direct ELISAs, less than 5% cross-reactivity with recombinant human (rh) IL-1 beta and rhIL-1F6 is observed and less than 1% cross-reactivity with rhIL-1 alpha and rhIL-18 is observed. Reconstitute at 0.2 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.

IL-36B is is a member of the interleukin 1 cytokine family whose gene and eight other interleukin 1 family genes form a cytokine gene cluster on chromosome 2. IL-36B is thought to activate the NF-kappaB pathway through IL-1 receptor family members IL-1RL2 and IL-1RAcP. Like the related proteins IL-36A and IL-36G, IL-36B requires post-translational processing for full agonist activity, but the cleavage mechanism is currently unknown. The IL-36 cytokines have been suggested to amplify Th1 responses by enhancing proliferation and Th1 polarization of naive CD4+ T cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL36B
Applications Neutralization
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene IL36B
Gene Accession No. Q9NZH7
Gene Alias 2310043N20Rik; family of interleukin 1-eta; FIL1; FIL1 eta; FIL1-(ETA); Fil1e; FIL1H; FILI-(ETA); IL-1 eta; IL1-ETA; IL1F8; IL-1F8; IL1F8 (Canonical product IL-1F8a); IL-1F8 (FIL1-eta); IL1H2; IL-1H2; Il36b; Interleukin; interleukin 1 family, member 8; interleukin 1 family, member 8 (eta); interleukin 1, eta; interleukin 36, beta; interleukin-1 eta; Interleukin-1 family member 8; interleukin-1 homolog 2; Interleukin-1 Superfamily e; interleukin-36 beta
Gene Symbols IL36B
Host Species Goat
Immunogen E. coli-derived recombinant human IL-36 beta/IL-1F8 Met1-Glu157.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27177
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.