Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IL36B Recombinant Rabbit Monoclonal Antibody (025)
GREENER_CHOICE

Catalog No. PIMA530769
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA530769 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA530769 Supplier Invitrogen™ Supplier No. MA530769
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Mouse IL36B/IL1F8.

IL-36B is is a member of the interleukin 1 cytokine family whose gene and eight other interleukin 1 family genes form a cytokine gene cluster on chromosome 2. IL-36B is thought to activate the NF-kappaB pathway through IL-1 receptor family members IL-1RL2 and IL-1RAcP. Like the related proteins IL-36A and IL-36G, IL-36B requires post-translational processing for full agonist activity, but the cleavage mechanism is currently unknown. The IL-36 cytokines have been suggested to amplify Th1 responses by enhancing proliferation and Th1 polarization of naive CD4+ T cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL36B
Applications ELISA
Classification Recombinant Monoclonal
Clone 025
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Il1f8
Gene Accession No. Q9D6Z6
Gene Alias 2310043N20Rik; family of interleukin 1-eta; FIL1; FIL1 eta; FIL1-(ETA); Fil1e; FIL1H; FILI-(ETA); IL-1 eta; IL1-ETA; IL1F8; IL-1F8; IL1F8 (Canonical product IL-1F8a); IL-1F8 (FIL1-eta); IL1H2; IL-1H2; Il36b; Interleukin; interleukin 1 family, member 8; interleukin 1 family, member 8 (eta); interleukin 1, eta; interleukin 36, beta; interleukin-1 eta; Interleukin-1 family member 8; interleukin-1 homolog 2; Interleukin-1 Superfamily e; interleukin-36 beta
Gene Symbols Il1f8
Host Species Rabbit
Immunogen Recombinant Mouse IL36B/IL1F8 protein (Met1-Lys183).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 69677
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.