Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL37 Mouse anti-Human, Clone: 7F1A1, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | IL37 |
---|---|
Clone | 7F1A1 |
Concentration | 1.5 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6391
![]() ![]() |
Proteintech Group Inc
602961IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
The protein encoded by this gene is a member of the interleukin 1 cytokine family. This cytokine can bind to, and may be a ligand for interleukin 18 receptor (IL18R1/IL-1Rrp). This cytokine also binds to interleukin 18 binding protein (IL18BP), an inhibitory binding protein of interleukin 18 (IL18), and subsequently forms a complex with IL18 receptor beta subunit, and through which it inhibits the activity of IL18. This gene along with eight other interleukin 1 family genes form a cytokine gene cluster on chromosome 2. Five alternatively spliced transcript variants encoding distinct isoforms have been reported.Specifications
IL37 | |
1.5 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
FIL1, FIL1 zeta, FIL1(ZETA), FIL1Z, IL 1 zeta, IL 1F7, IL 1H4, IL 1RP1, IL 1X, IL1F7, IL1H4, IL1RP1, IL37, Interleukin 1 family member 7, Interleukin 1 homolog 4, Interleukin 1 related protein, Interleukin 1 zeta | |
IL37 | |
IgG2a | |
Protein G | |
Antibody |
7F1A1 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human | |
Q9NZH6 | |
27178 | |
IL37 Fusion Protein Ag2441 | |
Primary | |
-20°C | |
IL37 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title