Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL3RA Mouse anti-Human, Clone: 10B8E7, Abnova™

Catalog No. 89934369 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-934-369 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-934-369 Supplier Abnova Corporation Supplier No. MAB12487
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant human IL3RA.

The protein encoded by this gene is an interleukin 3 specific subunit of a heterodimeric cytokine receptor. The receptor is comprised of a ligand specific alpha subunit and a signal transducing beta subunit shared by the receptors for interleukin 3 (IL3), colony stimulating factor 2 (CSF2/GM-CSF), and interleukin 5 (IL5). The binding of this protein to IL3 depends on the beta subunit. The beta subunit is activated by the ligand binding, and is required for the biological activities of IL3. This gene and the gene encoding the colony stimulating factor 2 receptor alpha chain (CSF2RA) form a cytokine receptor gene cluster in a X-Y pseudoautosomal region on chromosomes X or Y. [provided by RefSeq]

Specifications

Antigen IL3RA
Applications ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone 10B8E7
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant human IL3RA.
Dilution ELISA (1:10000) Flow Cytometry Immunocytochemistry (1:200-1:100) Immunofluorescence Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) Western Blot (1:500-1:2000) The optimal working dilution should be determ
Formulation In ascites (0.03% sodium azide)
Gene Alias CD123/IL3R/IL3RAY/IL3RX/IL3RY/MGC34174/hIL-3Ra
Gene Symbols IL3RA
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 200-305 of human IL3RA.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3563
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.