Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL3RA Rabbit anti-Human, Unconjugated, Clone: IL3RA/2947R, Abnova™

Catalog No. 89337883 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-337-883 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-337-883 Supplier Abnova Corporation Supplier No. RAB00458
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody.

IL3RA Monoclonal antibody specifically detects IL3RA in Human samples. It is validated for IHC-P.

Specifications

Antigen IL3RA
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone IL3RA/2947R
Conjugate Unconjugated
Description interleukin 3 receptor, alpha (low affinity)
Gene IL3RA
Gene Alias CD123, IL3R, IL3RAY, IL3RX, IL3RY, MGC34174, hIL-3Ra
Host Species Rabbit
Immunogen Original antibody is raised against recombinant protein corresponding to amino acid 26-171 of human IL3RA
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 3563
Target Species Human
Content And Storage Store at -20 to -80°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.