Learn More
Invitrogen™ IL4R Recombinant Rabbit Monoclonal Antibody (002), APC

Description
Preparation: This antibody was obtained from a rabbit immunized with purified, recombinant Rat IL4R (Met1-Arg232) and conjugated with APC under optimum conditions, the unreacted APC was removed. Storage: This antibody can be stored at 2C-8C for twelve months without detectable loss of activity. Protected from prolonged exposure to light. Do not freeze!.
Specifications
Specifications
| Antigen | IL4R |
| Applications | Flow Cytometry |
| Classification | Recombinant Monoclonal |
| Clone | 002 |
| Concentration | 100 μg/mL |
| Conjugate | APC |
| Formulation | PBS with 0.5% BSA and 0.03% Proclin 300 |
| Gene | IL4R |
| Gene Accession No. | Q63257 |
| Gene Alias | 582J2.1; CD124; il-4 receptor; IL-4 receptor alpha chain; IL-4 receptor subunit alpha; IL-4-binding protein; IL4-BP; Il4r; IL4R nirs variant 1; IL-4R subunit alpha; IL4RA; IL-4RA; IL-4R-alpha; interleukin 4 receptor; interleukin 4 receptor, alpha; interleukin-4 receptor alpha chain; interleukin-4 receptor subunit alpha; Soluble IL-4 receptor subunit alpha; Soluble interleukin-4 receptor subunit alpha |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.