Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IL5RA Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5121210
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121210 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5121210 Supplier Invitrogen™ Supplier No. PA5121210
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: HL-60, B cells, U-87MG, Mouse liver, Mouse lung, Rat lung. The target is usually found in the following locations: Membrane, Single-pass type I membrane protein. Immunogen sequence: ADLLPDEKIS LLPPVNFTIK VTGLAQVLLQ WKPNPDQEQR NVNLEYQVKI NAPKEDDYET RITESKCVTI LHKGFSASVR TILQNDHSLL ASSWASAELH APPGSPGTSI VNLTCTTNTT EDNYSRLRSY QVSLHCTWLV GTDAPEDTQY FLYYRYGSWT EECQEYSKDT LGRNIACWFP RTFILSKGRD WLAVLVNGSS KHSAIRPFDQ LFALHAIDQI NPPLNVTAEI EGTRLSIQWE KPVSAFPIHC FDYEVKIHNT RNGYLQIEKL MTNAFISIID DLSKYDVQVR AAVSSMCREA GLWSEWSQPI YVGFSR.

Interleukin 5 (IL-5) is a soluble T cell-derived factor, also known as T cell replacing factor (TRF), that induces B cell and eosinophil growth and differentiation. IL-5 exerts its biological activity through the IL-5 receptor (IL-5R), which is composed of two chains: an alpha chain that binds IL-5 with low affinity and a beta chain that does not bind IL-5, but together with the IL-5 alpha chain, constitutes the high affinity IL-5 receptor. The cytoplasmic domain of both the alpha and beta chains is essential for signal transduction. Specifically, the membrane-proximal proline-rich sequence of the cytoplasmic domain of the IL-5R receptor alpha subunit, IL-5Ralpha, is critical for the IL-5 induced proliferative response, expression of nuclear proto-oncogenes and tyrosine phosphorylation of cellular proteins, such as JAK1 and JAK2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL5RA
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.54 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene IL5RA
Gene Accession No. P21183, Q01344
Gene Alias CD125; CD125 antigen; CDw125; HSIL5R3; IL-5 receptor alpha chain; IL-5 receptor subunit alpha; Il5r; IL-5R subunit alpha; Il5ra; IL-5RA; IL-5R-alpha; interleukin 5 receptor subunit alpha; interleukin 5 receptor type 3; interleukin 5 receptor, alpha; interleukin-5 receptor alpha chain; interleukin-5 receptor subunit alpha
Gene Symbols IL5RA
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 20-335 of human IL5RA (NP_783855.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114103, 16192, 3568
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.