Learn More
Invitrogen™ IL5RA Polyclonal Antibody
Goat Polyclonal Antibody
Supplier: Invitrogen™ PA547750
Description
In direct ELISAs, approximately 10% cross-reactivity with recombinant human (rh) IL-5 R alpha is observed and less than 1% cross-reactivity with recombinant mouse (rm) IL-4 R, rhIL-9 R, rhIL-13 R alpha 1, and rhIL-13 R alpha 2 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
Interleukin 5 (IL-5) is a soluble T cell-derived factor, also known as T cell replacing factor (TRF), that induces B cell and eosinophil growth and differentiation. IL-5 exerts its biological activity through the IL-5 receptor (IL-5R), which is composed of two chains: an alpha chain that binds IL-5 with low affinity and a beta chain that does not bind IL-5, but together with the IL-5 alpha chain, constitutes the high affinity IL-5 receptor. The cytoplasmic domain of both the alpha and beta chains is essential for signal transduction. Specifically, the membrane-proximal proline-rich sequence of the cytoplasmic domain of the IL-5R receptor alpha subunit, IL-5Ralpha, is critical for the IL-5 induced proliferative response, expression of nuclear proto-oncogenes and tyrosine phosphorylation of cellular proteins, such as JAK1 and JAK2.
Specifications
IL5RA | |
Polyclonal | |
Unconjugated | |
IL5RA | |
CD125; CD125 antigen; CDw125; HSIL5R3; IL-5 receptor alpha chain; IL-5 receptor subunit alpha; Il5r; IL-5R subunit alpha; Il5ra; IL-5RA; IL-5R-alpha; interleukin 5 receptor subunit alpha; interleukin 5 receptor type 3; interleukin 5 receptor, alpha; interleukin-5 receptor alpha chain; interleukin-5 receptor subunit alpha | |
Goat | |
Antigen Affinity Chromatography | |
RUO | |
16192 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
P21183 | |
IL5RA | |
Mouse myeloma cell line NS0-derived recombinant mouse IL-5 R alpha/CD125 Asp18-His339. | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.