Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL6 Monoclonal Antibody (OTI12C5), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene CF813838


Description
IL6 Monoclonal antibody specifically detects IL6 in Human samples. It is validated for ELISASpecifications
IL6 | |
Monoclonal | |
See Label | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P05231 | |
Mouse | |
Protein A/G | |
RUO | |
3569 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG1 |
ELISA | |
OTI12C5 | |
Unconjugated | |
Il6 | |
B-cell differentiation factor; B-cell hybridoma growth factor; B-cell stimulatory factor 2; BSF2; BSF-2; CDF; CHIL-6; CTL differentiation factor; cytokine; HGF; H-IL-6; HSF; hybridoma growth factor; Ifnb2; IFN-beta-2; Il6; IL-6; ILg6; ILN; interferon beta-2; Interleukin; interleukin 6; Interleukin 6 (interferon, beta 2); interleukin 6 precursor; interleukin BSF-2; interleukin HP-1; Interleukin6; Interleukin-6; interleukin-6 precursor; interleukin-6 protein; M-IL-6; prointerleukin 6; R-IL-6 | |
Full length human recombinant protein of human IL-6 (NP_000591) produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Lyophilized |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction