Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL6R Antibody (B-F19), Allophycocyanin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP318095APC
Description
IL6R Monoclonal antibody specifically detects IL6R in Human samples. It is validated for Flow Cytometry, ELISA, ImmunohistochemistrySpecifications
IL6R | |
Monoclonal | |
APC | |
CD126, CD126 antigen, gp80, IL-6 receptor subunit alpha, IL-6R 1, IL-6R subunit alpha, IL-6R-1, IL6RA, IL-6RA, IL-6R-alpha, interleukin 6 receptor, interleukin 6 receptor alpha subunit, interleukin-6 receptor subunit alpha, Membrane glycoprotein 80, MGC104991 | |
gp80 transfected CHO cell line. Hybridoma: Myeloma X63/AG.8653 x Balb/c spleen cells | |
0.1 mL | |
Cancer, Cytokine Research, Immunology, Innate Immunity | |
3570 | |
Store at 4°C in the dark. | |
IgG1 κ |
Flow Cytometry, ELISA, Immunohistochemistry | |
B-F19 | |
PBS | |
Mouse | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction