Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL6R Antibody (D7715A7), Alexa Fluor™ 750, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP312009AF750
Description
IL6R Monoclonal antibody specifically detects IL6R in Mouse samples. It is validated for Flow Cytometry, ImmunofluorescenceSpecifications
IL6R | |
Monoclonal | |
Alexa Fluor 750 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Flow Cytometry, Immunofluorescence | |
D7715A7 | |
Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
CD126, CD126 antigen, gp80, IL-6 receptor subunit alpha, IL-6R 1, IL-6R subunit alpha, IL-6R-1, IL6RA, IL-6RA, IL-6R-alpha, interleukin 6 receptor, interleukin 6 receptor alpha subunit, interleukin-6 receptor subunit alpha, Membrane glycoprotein 80, MGC104991 | |
This antibody was raised by immunising Lewis rats with OKT4 cells. | |
0.1 mL | |
Cancer, Cytokine Research, Immunology, Innate Immunity | |
3570 | |
Store at 4C in the dark. | |
IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction