Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL6R Antibody (OTI2F4), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271029F
Description
IL6R Monoclonal specifically detects IL6R in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
IL6R | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
IL6R | |
Full length recombinant protein of human IL6R (NP_000556) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI2F4 | |
Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
CD126, CD126 antigen, gp80, IL-6 receptor subunit alpha, IL-6R 1, IL-6R subunit alpha, IL-6R-1, IL6RA, IL-6RA, IL-6R-alpha, interleukin 6 receptor, interleukin 6 receptor alpha subunit, interleukin-6 receptor subunit alpha, Membrane glycoprotein 80, MGC104991 | |
Mouse | |
Affinity Purified | |
Cancer, Cytokine Research, Immunology, Innate Immunity | |
3570.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction