Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL6R Antibody (RM0114-14F33), FITC, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP122492F
Description
IL6R Monoclonal specifically detects IL6R in Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
IL6R | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
IL6R | |
Mouse recombinant protein of the extracellular domain of IL-6 R | |
0.1 mL | |
Primary | |
This was produced from a hybridoma (mouse myeloma fused with spleen cells from a rat immunized with mouse recombinant protein of the extracellular domain of IL-6 R. | |
Store at 4C in the dark. | |
IgG2 |
Immunohistochemistry, Immunohistochemistry (Frozen) | |
RM0114-14F33 | |
Immunohistochemistry, Immunohistochemistry-Frozen | |
CD126, CD126 antigen, gp80, IL-6 receptor subunit alpha, IL-6R 1, IL-6R subunit alpha, IL-6R-1, IL6RA, IL-6RA, IL-6R-alpha, interleukin 6 receptor, interleukin 6 receptor alpha subunit, interleukin-6 receptor subunit alpha, Membrane glycoprotein 80, MGC104991 | |
Rat | |
Protein A or G purified | |
Cancer, Cytokine Research, Immunology, Innate Immunity | |
3570 | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction