Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL6R Mouse anti-Human, Clone: OTI2F4, liquid, TrueMAB™
SDP

Catalog No. 501676809
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-167-6809 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-167-6809 Supplier OriGene Supplier No. TA506859
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IL6R (interleukin-6 receptor subunit alpha) is a part of the receptor for interleukin 6. The IL-6 receptor complex is composed of two membrane glycoproteins: the low affinity receptor and the signal transducing component. Dysregulated production of IL6 and this receptor are implicated in the pathogenesis of many diseases, such as multiple myeloma, autoimmune diseases and prostate cancer.

Specifications

Antigen IL6R
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI2F4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene IL6R
Gene Accession No. P08887
Gene Alias CD126, IL-6R-1, IL-6RA, IL6Q, IL6RA, IL6RQ, gp80
Gene Symbols IL6R
Host Species Mouse
Immunogen Full length human recombit protein of human IL6R produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3570
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.