Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL7R Mouse anti-Human, Unconjugated, Clone: IL7R/2751, Abnova™

Catalog No. 89337594 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-337-594 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-337-594 Supplier Abnova Corporation Supplier No. MAB21101
Add to Cart
Add to Cart

Mouse Monoclonal Antibody.

IL7R Monoclonal antibody specifically detects IL7R in Human samples. It is validated for IHC-P.

Specifications

Antigen IL7R
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone IL7R/2751
Conjugate Unconjugated
Description interleukin 7 receptor
Gene IL7R
Gene Alias CD127, CDW127, IL-7R-alpha, IL7RA, ILRA
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acid 21-236 of human IL7R
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 3575
Target Species Human
Content And Storage Store at 2 to 8°C
Form Liquid
Isotype IgG2c κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.