Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IL9R alpha Monoclonal Antibody (AH9R7)
GREENER_CHOICE

Catalog No. PIMA528556
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA528556 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA528556 Supplier Invitrogen™ Supplier No. MA528556
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody recognizes an extracellular epitope of CD129 / IL-9R alpha, a 57 kDa type I transmembrane glycoprotein expressed at low levels by lymphocytes, blood cell progenitors, eosinophils, mast cells, epithelial cells, muscle cells and neurons. Flow cytometry: It is recommended to use bright fluorochromes or signal multiplying detection; Functional Application: Blocking.

The protein encoded by this gene is a cytokine receptor that specifically mediates the biological effects of interleukin 9 (IL9). The functional IL9 receptor complex requires this protein as well as the interleukin 2 receptor, gamma (IL2RG), a common gamma subunit shared by the receptors of many different cytokines. The ligand binding of this receptor leads to the activation of various JAK kinases and STAT proteins, which connect to different biologic responses. This gene is located at the pseudoautosomal regions of X and Y chromosomes. Genetic studies suggested an association of this gene with the development of asthma. Multiple pseudogenes on chromosome 9, 10, 16, and 18 have been described. Alternatively spliced transcript variants have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL9R alpha
Applications ELISA, Flow Cytometry, Functional Assay
Classification Monoclonal
Clone AH9R7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene IL9R
Gene Accession No. Q01113
Gene Alias CD129; IL-9 receptor; IL9R; IL-9R; interleukin 9 receptor; interleukin-9 receptor
Gene Symbols IL9R
Host Species Mouse
Immunogen Human CD129-transfected cell line.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3581
Target Species Human
Content And Storage 4°C, do not freeze
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.