Learn More
Invitrogen™ IL9R alpha Monoclonal Antibody (AH9R7)

Description
This antibody recognizes an extracellular epitope of CD129 / IL-9R alpha, a 57 kDa type I transmembrane glycoprotein expressed at low levels by lymphocytes, blood cell progenitors, eosinophils, mast cells, epithelial cells, muscle cells and neurons. Flow cytometry: It is recommended to use bright fluorochromes or signal multiplying detection.
Specifications
Specifications
| Antigen | IL9R alpha |
| Applications | ELISA, Flow Cytometry |
| Classification | Monoclonal |
| Clone | AH9R7 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 15mM sodium azide; pH 7.4 |
| Gene | IL9R |
| Gene Accession No. | Q01113 |
| Gene Alias | CD129; IL-9 receptor; IL9R; IL-9R; interleukin 9 receptor; interleukin-9 receptor |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.