Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IL9R Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584592
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584592 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584592 Supplier Invitrogen™ Supplier No. PA584592
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LTNNILRIDC HWSAPELGQG SSPWLLFTSN QAPGGTHKCI LRGSECTVVL PPEAVLVPSD NFTITFHHCM SGREQVSLVD PEYLPRR Highest antigen sequence indentity to the following orthologs: Mouse - 67%, Rat - 61%.

Human Interleukin 9 (IL9) was first identified based on its proliferative activity on a human myeloid leukemic cell line. Human IL9 stimulates the proliferation of the human megakaryocytic leukemic cell line M07e. Human IL9 supports erythroid colony formation and synergizes with IL4 in the production of IgE and IgG. The interleukin 9 receptor is a member of the hematopoietin receptor superfamily. The cDNAs encoding mouse and human IL9 receptors have been isolated. The deduced mouse and human transmembrane proteins, sharing 53% amino acid sequence identity, contain 468 and 533 amino acid residues, respectively. A number of isoforms of IL9R, including a putative soluble form, have also been identified. The IL9 receptor is expressed in a variety of hematopoietic cells including T cells, neutrophils, mast cells, and macrophages.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL9R
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene IL9R
Gene Accession No. Q01113
Gene Alias CD129; IL-9 receptor; Il9r; IL-9R; interleukin 9 receptor; interleukin-9 receptor
Gene Symbols IL9R
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human IL9R. Recombinant protein control fragment (Product #RP-108487).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3581
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.