Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ILDR1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570149
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570149 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570149 Supplier Invitrogen™ Supplier No. PA570149
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Guinea Pig: 93%; Horse: 86%; Human: 100%; Mouse: 86%; Rat: 79%.

This gene encodes a protein that contains an immunoglobulin-like domain. The encoded protein may function as a multimeric receptor at the cell surface. The expression of this gene may be a diagnostic marker for cancer progression. Alternatively spliced transcript variants encoding multiple protein isoforms have been observed for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ILDR1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ILDR1
Gene Accession No. Q86SU0
Gene Alias AU041483; AU044638; DFNB42; ILDR1; ILDR1alpha; ILDR1alpha'; ILDR1beta; immunoglobulin like domain containing receptor 1; immunoglobulin-like domain containing receptor 1; immunoglobulin-like domain-containing receptor 1; immunoglobulin-like domain-containing receptor 1 alpha; immunoglobulin-like domain-containing receptor 1 beta; RGD1307792
Gene Symbols ILDR1
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human ILDR1.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 286676
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.