Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ILDR1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA553755
Description
Immunogen sequence: LEYLEKELRN LNLAQPLPPD LKGRFGHPCS MLSSLGSEVV ERRIIHLPPL IRDLSSSRRT SDSLHQQWLT PIPSRPWDLR EGRSHHHYPD FHQELQDRGP KSWALERREL DPSWSGRHRS SRLNGSPIHW SDRDSLSDVP S Highest antigen sequence identity to the following orthologs: Mouse - 65%, Rat - 68%.
This gene encodes a protein that contains an immunoglobulin-like domain. The encoded protein may function as a multimeric receptor at the cell surface. The expression of this gene may be a diagnostic marker for cancer progression. Alternatively spliced transcript variants encoding multiple protein isoforms have been observed for this gene.
Specifications
ILDR1 | |
Polyclonal | |
Unconjugated | |
ILDR1 | |
AU041483; AU044638; DFNB42; ILDR1; ILDR1alpha; ILDR1alpha'; ILDR1beta; immunoglobulin like domain containing receptor 1; immunoglobulin-like domain containing receptor 1; immunoglobulin-like domain-containing receptor 1; immunoglobulin-like domain-containing receptor 1 alpha; immunoglobulin-like domain-containing receptor 1 beta; RGD1307792 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
286676 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q86SU0 | |
ILDR1 | |
Recombinant protein corresponding to Human ILDR1. Recombinant protein control fragment (Product #RP-90984). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction