Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ILF1 Antibody, Novus Biologicals™

Goat Polyclonal Antibody has been used in 2 publications
Supplier: Novus Biologicals NB1001285
Description
ILF1 Polyclonal specifically detects ILF1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Peptide ELISA.Specifications
ILF1 | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
Cellular transcription factor ILF-1, forkhead box K2, forkhead box protein K2, FOXK1, ILF-1, ILF1ILF, interleukin enhancer binding factor 1, Interleukin enhancer-binding factor 1 | |
Goat | |
69 kDa | |
0.1 mg | |
Immunology, Innate Immunity | |
3607 | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, ELISA | |
0.5 mg/mL | |
Western Blot 0.01 - 0.03 μg/mL, Flow Cytometry 10 μg/mL, Immunocytochemistry/Immunofluorescence 10 μg/mL, Peptide ELISA Detection limit 1:32000 | |
Q01167 | |
FOXK2 | |
Peptide with sequence C-TPPAAVREKGVQN corresponding to C-Terminus according to NP_004505.2. | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction