Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ILF1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582981
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582981 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582981 Supplier Invitrogen™ Supplier No. PA582981
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SSQPVLITVQ RQLPQAIKPV TYTVATPVTT STSQPPVVQT VHVVHQIPAV SVTSVAGLAP ANTYTVSGQA VVTPAAVLAP PKAEAQENGD HREVKVKVEP IPAIGHATLG TASRIIQTAQ TTPVQTVTIV QQAPLGQHQL PIK Highest antigen sequence indentity to the following orthologs: Rat - 64%, Mouse - 90%.

The protein encoded by this gene contains a fork head DNA binding domain. This protein can bind to the purine-rich motifs of the HIV long terminal repeat (LTR), and to the similar purine-rich motif in the interleukin 2 (IL2) promoter. It may be involved in the regulation of viral and cellular promoter elements.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ILF1
Applications ChIP Assay, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Foxk2
Gene Accession No. Q01167
Gene Alias 1110054H05Rik; 5730434B08Rik; 6230415M23Rik; cellular transcription factor ILF-1; forkhead box K2; forkhead box protein K2; FOXK1; foxk2; G/T-mismatch specific binding protein; ILF; ILF1; ILF-1; interleukin enhancer binding factor 1; interleukin enhancer-binding factor 1; nGTBP
Gene Symbols Foxk2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FOXK2. Recombinant protein control fragment (Product #RP-100721).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3607
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.