Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ILF1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5115419
Description
Antibody detects endogenous levels of total ILF1.
The protein encoded by this gene contains a fork head DNA binding domain. This protein can bind to the purine-rich motifs of the HIV long terminal repeat (LTR), and to the similar purine-rich motif in the interleukin 2 (IL2) promoter. It may be involved in the regulation of viral and cellular promoter elements.
Specifications
ILF1 | |
Polyclonal | |
Unconjugated | |
Foxk2 | |
1110054H05Rik; 5730434B08Rik; 6230415M23Rik; cellular transcription factor ILF-1; forkhead box K2; forkhead box protein K2; FOXK1; foxk2; G/T-mismatch specific binding protein; ILF; ILF1; ILF-1; interleukin enhancer binding factor 1; interleukin enhancer-binding factor 1; nGTBP | |
Rabbit | |
Affinity chromatography | |
RUO | |
303753, 3607, 68837 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide | |
Q01167, Q3UCQ1 | |
Foxk2 | |
A synthesized peptide derived from human FOXK2(Accession Q01167), corresponding to amino acid residues E628-N660. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction