Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ILF1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5119717
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5119717 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5119717 Supplier Invitrogen™ Supplier No. PA5119717
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive Control: 293 cell lysates, mouse lung tissue, mouse colon tissue, mouse brain tissue, SHSY5Y. Subcellular Location: Cytoplasm, Nucleus.

The protein encoded by this gene contains a fork head DNA binding domain. This protein can bind to the purine-rich motifs of the HIV long terminal repeat (LTR), and to the similar purine-rich motif in the interleukin 2 (IL2) promoter. It may be involved in the regulation of viral and cellular promoter elements.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ILF1
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
Gene Foxk2
Gene Accession No. Q01167, Q3UCQ1
Gene Alias 1110054H05Rik; 5730434B08Rik; 6230415M23Rik; cellular transcription factor ILF-1; forkhead box K2; forkhead box protein K2; FOXK1; foxk2; G/T-mismatch specific binding protein; ILF; ILF1; ILF-1; interleukin enhancer binding factor 1; interleukin enhancer-binding factor 1; nGTBP
Gene Symbols Foxk2
Host Species Rabbit
Immunogen Recombinant protein within mouse Foxk2 aa 1-350.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 303753, 3607, 68837
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.