Learn More
Invitrogen™ ILF2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA589159
Description
Immunogen sequence: MRGDRGRGRG GRFGSRGGPG GGFRPFVPHI PFDFYLCEMA FPRVKPAPDE TSFSEALLKR NQDLAPNSAE QASILSLVTK INNVIDNLIV APGTFEVQIE EVRQVGSYKK GTMTTGHNVA DLVVILKILP TLEAVAALGN KVVESLRAQD PSEVLTMLTN ETGFEISSSD ATVKILITTV PPNLRKLDPE LHLDIKVLQS ALAAIRHARW FEENASQSTV KVLIRLLKDL RIRFPGFEPL TPWILDLLGH YAVMNNPTRQ PLALNVAYRR CLQILAAGLF; Positive Samples: MCF-7, Raji; Cellular Location: Cytoplasm, Nucleus, nucleolus.
Nuclear factor of activated T-cells is a transcription factor required for T-cell expression of the interleukin 2 gene. NFAT binds to a sequence in the interleukin 2 gene enhancer known as the antigen receptor response element 2. In addition, NFAT can bind RNA and is an essential component for encapsidation and protein priming of hepatitis B viral polymerase. NFAT is a heterodimer of 45 kDa and 90 kDa proteins, the smaller of which is the product of this gene. The encoded protein binds strongly to the 90 kDa protein and stimulates its ability to enhance gene expression.
Specifications
ILF2 | |
Polyclonal | |
Unconjugated | |
ILF2 | |
6230405A16Rik; Ab1-143; ILF2; interleukin enhancer binding factor 2; interleukin enhancer binding factor 2, 45kD; interleukin enhancer binding factor 2, 45kDa; interleukin enhancer-binding factor 2; Interleukin enhancer-binding factor 2 homolog; liver regeneration-related protein LRRG031; MGC8391; Nf45; nuclear factor of activated T-cells 45 kDa; nuclear factor of activated T-cells, 45-kDa; PRO3063; TEG-267; testis expressed gene 267; Tex261; Tex267; Unknown (protein for MGC:134524); wu:fb34h01; zgc:73084; zgc:77276 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
3608, 67781 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.28 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q12905, Q9CXY6 | |
ILF2 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-280 of human ILF2 (NP_0045062). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.