Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ILF2 Rabbit anti-Human, Mouse, Rat, Clone: JU50-33, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP275539
Description
ILF2 Monoclonal specifically detects ILF2 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
ILF2 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:100-1:2000, Flow Cytometry 1:50-1:200, Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:50-1:200 | |
interleukin enhancer binding factor 2, 45kD, interleukin enhancer binding factor 2, 45kDa, interleukin enhancer-binding factor 2, MGC8391, NF45PRO3063, Nuclear factor of activated T-cells 45 kDa, nuclear factor of activated T-cells, 45-kDa | |
Rabbit | |
43 kDa | |
100 μL | |
3608.0 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
JU50-33 | |
Unconjugated | |
TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide | |
ILF2 | |
Synthetic peptide within Human ILF2 aa 245-294/ 390. (SwissProt: Q12905 Human; SwissProt: Q9CXY6 Mouse; SwissProt: Q7TP98 Rat) | |
Protein A purified | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction